accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaSH

CAS RN 207683-19-0

IUPAC Name: sodium hydrate sulfanide
Synonyms: sodium hydrate sulfanide
Molecular Weight (g/mol): 74.07
Molecular Formula: H3NaOS
InChi Key: ZNKXTIAQRUWLRL-UHFFFAOYSA-M
SMILES: O.[Na+].[SH-]
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Grade 
  • (4)
Percent Purity 
  • (4)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)
  • (4)