accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H2NH3CCH3NH2CH3HClHCl

CAS RN 207738-08-7

IUPAC Name: 3,3',5,5'-tetramethyl-[1,1'-biphenyl]-4,4'-diamine dihydrate dihydrochloride
Synonyms: tetramethylbenzidine dihydrate dihydrochloride
Molecular Weight (g/mol): 349.3
Molecular Formula: C16H26Cl2N2O2
InChi Key: CJRXJBONCVPXPJ-UHFFFAOYSA-N
SMILES: O.O.Cl.Cl.CC1=CC(=CC(C)=C1N)C1=CC(C)=C(N)C(C)=C1
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results