accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 20780-53-4

IUPAC Name: (2R)-2-phenyloxirane
Synonyms: R-styrene oxide
Molecular Weight (g/mol): 120.15
Molecular Formula: C8H8O
InChi Key: AWMVMTVKBNGEAK-QMMMGPOBSA-N
SMILES: C1O[C@@H]1C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (6)
Quantity 
  • (3)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (6)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)