accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 207801-31-8

IUPAC Name: zinc(2+) hydrate ditrifluoroacetate
Synonyms: zinc(2+) hydrate ditrifluoroacetate
Molecular Weight (g/mol): 309.43
Molecular Formula: C4H2F6O5Zn
InChi Key: VUNLDTPRUICHRY-UHFFFAOYSA-L
SMILES: O.[Zn++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)