accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FNHSNH2F

CAS RN 207981-44-0

IUPAC Name: (2,5-difluorophenyl)thiourea
Synonyms: 2,5-difluorophenylthiourea
Molecular Weight (g/mol): 188.2
Molecular Formula: C7H6F2N2S
InChi Key: JBWSNSWRTOFDBM-UHFFFAOYSA-N
SMILES: NC(=S)NC1=C(F)C=CC(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results