accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOsOOO

CAS RN 20816-12-0

IUPAC Name: tetraoxoosmium
Synonyms: osmium tetroxide
Molecular Weight (g/mol): 254.23
Molecular Formula: O4Os
InChi Key: VUVGYHUDAICLFK-UHFFFAOYSA-N
SMILES: O=[Os](=O)(=O)=O
Quantity 
  • (1)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (4)
  • (1)
Physical Form 
  • (3)
  • (5)
  • (3)
  • (6)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (5)
  • (3)
  • (3)
Grade 
  • (3)
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (7)
  • (3)
  • (11)
  • (7)
  • (2)
  • (1)
  • (4)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)
  • (1)
  • (4)
  • (1)
  • (3)
  • (5)
  • (3)
  • (6)
  • (2)
  • (2)
  • (2)
  • (5)
  • (3)
  • (3)
  • (3)
  • (5)