accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOFFF

CAS RN 208242-25-5

IUPAC Name: methyl 1-(trifluoromethyl)cyclopropane-1-carboxylate
Synonyms: methyl 1-(trifluoromethyl)cyclopropane-1-carboxylate
Molecular Weight (g/mol): 168.12
Molecular Formula: C6H7F3O2
InChi Key: PAWUSHZXEZPDBE-UHFFFAOYSA-N
SMILES: COC(=O)C1(CC1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results