accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 208255-80-5

CH3ONHOOCH3H3CCH3HNOFF

CAS RN 208255-80-5

IUPAC Name: tert-butyl 2-{2-[2-(3,5-difluorophenyl)acetamido]propanamido}-2-phenylacetate
Synonyms: tert-butyl 2-{2-[2-(3,5-difluorophenyl)acetamido]propanamido}-2-phenylacetate
Molecular Weight (g/mol): 432.47
Molecular Formula: C23H26F2N2O4
InChi Key: DWJXYEABWRJFSP-UHFFFAOYNA-N
SMILES: CC(NC(=O)CC1=CC(F)=CC(F)=C1)C(=O)NC(C(=O)OC(C)(C)C)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)