accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3BrOHH3CH3CCH3

CAS RN 20834-61-1

IUPAC Name: 2-bromo-4,6-di-tert-butylphenol
Synonyms: 2-bromo-4,6-di-tert-butylphenol
Molecular Weight (g/mol): 285.23
Molecular Formula: C14H21BrO
InChi Key: DIWZVAHZEOFSLS-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(Br)=C(O)C(=C1)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results