accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COBOHHOH3C

CAS RN 208399-66-0

IUPAC Name: (4-methoxy-2-methylphenyl)boronic acid
Synonyms: 4-methoxy-2-methylphenylboronic acid
Molecular Weight (g/mol): 165.98
Molecular Formula: C8H11BO3
InChi Key: AMSQNQJCBXQYEX-UHFFFAOYSA-N
SMILES: COC1=CC=C(B(O)O)C(C)=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)