accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 20859-02-3

IUPAC Name: 2-amino-3,3-dimethylbutanoic acid
Synonyms: 2-amino-3,3-dimethylbutanoic acid
Molecular Weight (g/mol): 131.18
Molecular Formula: C6H13NO2
InChi Key: NPDBDJFLKKQMCM-UHFFFAOYNA-N
SMILES: CC(C)(C)C(N)C(O)=O
Molecular Weight (g/mol) 
  • (1)
  • (6)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (1)
  • (6)
  • (2)
  • (2)
  • (2)
  • (1)