accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2096330-26-4

BOOH3CCH3H3CH3COHNO

CAS RN 2096330-26-4

IUPAC Name: N-(benzyloxy)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Synonyms: N-(benzyloxy)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide
Molecular Weight (g/mol): 353.23
Molecular Formula: C20H24BNO4
InChi Key: XECXLZBBBRYLFW-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(C=C1)C(=O)NOCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results