accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BSOONCH3HOOH

CAS RN 2096336-05-7

IUPAC Name: {4-[methyl(phenyl)sulfamoyl]phenyl}boronic acid
Synonyms: 4-[methyl(phenyl)sulfamoyl]phenylboronic acid
Molecular Weight (g/mol): 291.13
Molecular Formula: C13H14BNO4S
InChi Key: PUWVRBKKJNKHLI-UHFFFAOYSA-N
SMILES: CN(C1=CC=CC=C1)S(=O)(=O)C1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results