accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2096338-17-7

BOHNH3CClHOOH

CAS RN 2096338-17-7

IUPAC Name: [2-chloro-5-(methylcarbamoyl)phenyl]boronic acid
Synonyms: 2-chloro-5-(methylcarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 213.42
Molecular Formula: C8H9BClNO3
InChi Key: YTEOTIIPHSQWBH-UHFFFAOYSA-N
SMILES: CNC(=O)C1=CC(B(O)O)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results