CAS RN 2096341-40-9
IUPAC Name:
{2-fluoro-5-[(2-nitrophenyl)carbamoyl]phenyl}boronic acid
Synonyms:
2-fluoro-5-[(2-nitrophenyl)carbamoyl]phenylboronic acid
Molecular Weight (g/mol):
304.04
Molecular Formula:
C13H10BFN2O5
InChi Key:
LZCKVRHHMFHKQO-UHFFFAOYSA-N
SMILES:
OB(O)C1=C(F)C=CC(=C1)C(=O)NC1=CC=CC=C1[N+]([O-])=O
1
–
1
of
1
results
1
2-Fluoro-5-(2-nitrophenylcarbamoyl)benzeneboronic acid, 97%, Thermo Scientific™
CAS: 2096341-40-9 Molecular Formula: C13H10BFN2O5 Molecular Weight (g/mol): 304.04 MDL Number: MFCD20265327 InChI Key: LZCKVRHHMFHKQO-UHFFFAOYSA-N Synonym: 2-fluoro-5-2-nitrophenylcarbamoyl benzeneboronic acid PubChem CID: 73995936