accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 20980-22-7

IUPAC Name: 2-(piperazin-1-yl)pyrimidine
Synonyms: pyrimidinylpiperazine
Molecular Weight (g/mol): 164.21
Molecular Formula: C8H12N4
InChi Key: MRBFGEHILMYPTF-UHFFFAOYSA-N
SMILES: C1CN(CCN1)C1=NC=CC=N1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)