CAS RN 209810-38-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(R/S)-Colchicine, TRC
CAS: 209810-38-8 Molecular Formula: C22 H25 N O6 Molecular Weight (g/mol): 399.44 Synonym: N-[5,6,7,9-Tetrahydro-1,2,3,10-tetramethoxy-9-oxobenzo[a]heptalen-7-yl]acetamide,(+/-)-Colchicine,(7S,12aS)-Colchicine,(-)-N-[(7S,12aSa)-1,2,3,10-Tetramethoxy-9-oxo-5,6,7,9-tetrahydrobenzo[a]heptalen-7-yl]acetamide (Conformational Isomer),Acetamide, N-[(7S,12aS)-5,6,7,9-tetrahydro-1,2,3,10-tetramethoxy-9-oxobenzo[a]heptalen-7-yl]- (9CI),N-[(7S,12aS IUPAC Name: N-(1,2,3,10-tetramethoxy-9-oxo-6,7-dihydro-5H-benzo[a]heptalen-7-yl)acetamide SMILES: COC1=CC=C2C(=CC1=O)[C@H](CCc3cc(OC)c(OC)c(OC)c23)NC(=O)C