accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2102510-60-9

IUPAC Name: 3-ethyl-5-{[7-(15-{7-[(3-ethyl-4-oxo-2-sulfanylidene-1,3-thiazolidin-5-ylidene)methyl]-2,1,3-benzothiadiazol-4-yl}-9,9,18,18-tetrakis(2-ethylhexyl)-5,14-dithiapentacyclo[10.6.0.0³,¹⁰.0⁴,⁸.0¹³,¹⁷]octadeca-1(12),2,4(8),6,10,13(17),15-heptaen-6-yl)-2,1,3-benzothiadiazol-4-yl]methylidene}-2-sulfanylidene-1,3-thiazolidin-4-one
Synonyms: 3-ethyl-5-{[7-(15-{7-[(3-ethyl-4-oxo-2-sulfanylidene-1,3-thiazolidin-5-ylidene)methyl]-2,1,3-benzothiadiazol-4-yl}-9,9,18,18-tetrakis(2-ethylhexyl)-5,14-dithiapentacyclo[10.6.0.0³,¹⁰.0⁴,⁸.0¹³,¹⁷]octadeca-1(12),2,4(8),6,10,13(17),15-heptaen-6-yl)-2,1,3-benzothiadiazol-4-yl]methylidene}-2-sulfanylidene-1,3-thiazolidin-4-one
Molecular Weight (g/mol): 1326.02
Molecular Formula: C72H88N6O2S8
InChi Key: HKJHYTKBDDVGRK-UHFFFAOYNA-N
SMILES: CCCCC(CC)CC1(CC(CC)CCCC)C2=C(SC(=C2)C2=CC=C(C=C3SC(=S)N(CC)C3=O)C3=NSN=C23)C2=CC3=C(C=C12)C1=C(C=C(S1)C1=CC=C(C=C2SC(=S)N(CC)C2=O)C2=NSN=C12)C3(CC(CC)CCCC)CC(CC)CCCC
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)