accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2103-91-5

IUPAC Name: 4-(4-methylphenyl)-1,3-thiazol-2-amine
Synonyms: 4-(4-methylphenyl)-1,3-thiazol-2-amine
Molecular Weight (g/mol): 190.26
Molecular Formula: C10H10N2S
InChi Key: ARLHWYFAPHJCJT-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1=CSC(N)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results