accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2SNCl

CAS RN 2103-99-3

IUPAC Name: 4-(4-chlorophenyl)-1,3-thiazol-2-amine
Synonyms: 4-(4-chlorophenyl)-1,3-thiazol-2-amine
Molecular Weight (g/mol): 210.68
Molecular Formula: C9H7ClN2S
InChi Key: DWGWNNCHJPKZNC-UHFFFAOYSA-N
SMILES: NC1=NC(=CS1)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results