accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 210532-25-5

IUPAC Name: 3,5-difluorobenzene-1-sulfonyl chloride
Synonyms: 3,5-difluorobenzenesulfonyl chloride
Molecular Weight (g/mol): 212.59
Molecular Formula: C6H3ClF2O2S
InChi Key: IIQKIICAOXAXEJ-UHFFFAOYSA-N
SMILES: FC1=CC(=CC(F)=C1)S(Cl)(=O)=O
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results