accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NH3CCH3OPOOONOO

CAS RN 21064-69-7

IUPAC Name: 4-nitrophenyl 2-(trimethylazaniumyl)ethyl phosphate
Synonyms: 4-nitrophenyl 2-(trimethylammonio)ethyl phosphate
Molecular Weight (g/mol): 304.24
Molecular Formula: C11H17N2O6P
InChi Key: NAIXASFEPQPICN-UHFFFAOYSA-N
SMILES: C[N+](C)(C)CCOP([O-])(=O)OC1=CC=C(C=C1)[N+]([O-])=O
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results