accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2107-77-9

IUPAC Name: 7,8-dihydroxy-4-methyl-2H-chromen-2-one
Synonyms: 7,8-dihydroxy-4-methylcoumarin
Molecular Weight (g/mol): 192.17
Molecular Formula: C10H8O4
InChi Key: NWQBYMPNIJXFNQ-UHFFFAOYSA-N
SMILES: CC1=CC(=O)OC2=C(O)C(O)=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results