accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

I

CAS RN 2113-51-1

IUPAC Name: 2-iodo-1,1'-biphenyl
Synonyms: 1,1'-biphenyl, 2-iodo-
Molecular Weight (g/mol): 280.11
Molecular Formula: C12H9I
InChi Key: QFUYDAGNUJWBSM-UHFFFAOYSA-N
SMILES: IC1=CC=CC=C1C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results