accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2117-12-6

IUPAC Name: 4-(trimethylsilyl)but-3-yn-1-ol
Synonyms: 4-(trimethylsilyl)but-3-yn-1-ol
Molecular Weight (g/mol): 142.27
Molecular Formula: C7H14OSi
InChi Key: VSTMVAFCUIGHQI-UHFFFAOYSA-N
SMILES: C[Si](C)(C)C#CCCO
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results