accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 211742-82-4

CH3ONHSOOHNOCH3OCH3

CAS RN 211742-82-4

IUPAC Name: N-{4-[(2,5-dimethoxyphenyl)sulfamoyl]phenyl}acetamide
Synonyms: N-{4-[(2,5-dimethoxyphenyl)sulfamoyl]phenyl}acetamide
Molecular Weight (g/mol): 350.39
Molecular Formula: C16H18N2O5S
InChi Key: VVYBOPRFGIFZSH-UHFFFAOYSA-N
SMILES: COC1=CC(NS(=O)(=O)C2=CC=C(NC(C)=O)C=C2)=C(OC)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results