accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNOO

CAS RN 21203-68-9

IUPAC Name: 2-methyl-5-nitropyridine
Synonyms: 2-methyl-5-nitropyridine
Molecular Weight (g/mol): 138.13
Molecular Formula: C6H6N2O2
InChi Key: USZINSZJSVMICC-UHFFFAOYSA-N
SMILES: CC1=NC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results