accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 21222-05-9

ONOONOO

CAS RN 21222-05-9

IUPAC Name: bis(3-nitrophenyl)methanone
Synonyms: bis(3-nitrophenyl)methanone
Molecular Weight (g/mol): 272.22
Molecular Formula: C13H8N2O5
InChi Key: BSDKBWGNIJMCID-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC=C1)C(=O)C1=CC(=CC=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results