accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FSO

CAS RN 21243-18-5

IUPAC Name: 6-fluoro-3,4-dihydro-2H-1-benzothiopyran-4-one
Synonyms: 6-fluoro-2,3-dihydro-1-benzothiopyran-4-one
Molecular Weight (g/mol): 182.21
Molecular Formula: C9H7FOS
InChi Key: NBBGHADNMPMHST-UHFFFAOYSA-N
SMILES: FC1=CC=C2SCCC(=O)C2=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)