accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 21256-18-8

IUPAC Name: 3-(4,5-diphenyl-1,3-oxazol-2-yl)propanoic acid
Synonyms: oxaprozin apotex brand of oxaprozin pharmacia brand of oxaprozin daypro rhoxalpharma brand of oxaprozin rhoxal-oxaprozin csc brand of oxaprozin Voir dayrun oxaprozina
Molecular Weight (g/mol): 293.32
Molecular Formula: C18H15NO3
InChi Key: OFPXSFXSNFPTHF-UHFFFAOYSA-N
SMILES: OC(=O)CCC1=NC(=C(O1)C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results