accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BClClClHOOH

CAS RN 212779-19-6

IUPAC Name: (2,3,5-trichlorophenyl)boronic acid
Synonyms: 2,3,5-trichlorophenylboronic acid
Molecular Weight (g/mol): 225.26
Molecular Formula: C6H4BCl3O2
InChi Key: OPBCCRZCYTUJMS-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(Cl)=CC(Cl)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results