accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2129-31-9

POOH

CAS RN 2129-31-9

IUPAC Name: 4-(diphenylphosphanyl)benzoic acid
Synonyms: 4-(diphenylphosphanyl)benzoic acid
Molecular Weight (g/mol): 306.3
Molecular Formula: C19H15O2P
InChi Key: GXMHDTPYKRTARV-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results