CAS RN 2131-59-1
IUPAC Name:
1-bromo-3-isothiocyanatobenzene
Synonyms:
1-bromo-3-isothiocyanatobenzene
Molecular Weight (g/mol):
214.08
Molecular Formula:
C7H4BrNS
InChi Key:
ZMGMGHNOACSMQN-UHFFFAOYSA-N
SMILES:
BrC1=CC=CC(=C1)N=C=S
1
–
2
of
2
results
1
3-Bromophenyl isothiocyanate, 97%
CAS: 2131-59-1 Molecular Formula: C7H4BrNS Molecular Weight (g/mol): 214.08 MDL Number: MFCD00004803 InChI Key: ZMGMGHNOACSMQN-UHFFFAOYSA-N Synonym: 3-bromophenyl isothiocyanate,m-bromophenyl isothiocyanate,benzene, 1-bromo-3-isothiocyanato,isothiocyanic acid, m-bromophenyl ester,3-bromophenylisothiocyanate,1-bromo-3-isothiocyanato-benzene,acmc-1chhg,m-bromphenylisothiocyanat,zmgmghnoacsmqn-uhfffaoysa,# PubChem CID: 16481 IUPAC Name: 1-bromo-3-isothiocyanatobenzene SMILES: C1=CC(=CC(=C1)Br)N=C=S