accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2131-61-5

IUPAC Name: 1-isothiocyanato-4-nitrobenzene
Synonyms: 4-nitrophenylisothiocyanate
Molecular Weight (g/mol): 180.18
Molecular Formula: C7H4N2O2S
InChi Key: NXHSSIGRWJENBH-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)N=C=S
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)