accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHONCS

CAS RN 2131-63-7

IUPAC Name: 3-isothiocyanatobenzoic acid
Synonyms: 3-isothiocyanatobenzoic acid
Molecular Weight (g/mol): 179.19
Molecular Formula: C8H5NO2S
InChi Key: PJRBPKOOGLKPFB-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=CC(=C1)N=C=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results