accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 2131-64-8

H3CNCH3NCS

CAS RN 2131-64-8

IUPAC Name: 4-isothiocyanato-N,N-dimethylaniline
Synonyms: 4-isothiocyanato-N,N-dimethylaniline
Molecular Weight (g/mol): 178.25
Molecular Formula: C9H10N2S
InChi Key: HRDJPEMAGYHSJR-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)N=C=S
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results