accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 21314-17-0

NNNOHN

CAS RN 21314-17-0

IUPAC Name: 1-phenyl-1H,2H,4H-pyrazolo[3,4-d]pyrimidin-4-one
Synonyms: 1-phenyl-2H-pyrazolo[3,4-d]pyrimidin-4-one
Molecular Weight (g/mol): 212.21
Molecular Formula: C11H8N4O
InChi Key: SVKPVSOBZKPDIY-UHFFFAOYSA-N
SMILES: O=C1N=CN=C2N(NC=C12)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results