accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

PbOOOCCCH3CH3H3CH3CCH3H3CH3CCH3H3CCH3CH3H3CHHO

CAS RN 21319-43-7

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: —
InChi Key: —
SMILES: —
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)