accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COSClOONOO

CAS RN 21320-91-2

IUPAC Name: 2-methoxy-4-nitrobenzene-1-sulfonyl chloride
Synonyms: 2-methoxy-4-nitrobenzenesulfonyl chloride
Molecular Weight (g/mol): 251.64
Molecular Formula: C7H6ClNO5S
InChi Key: QECYXMKYZQXEHM-UHFFFAOYSA-N
SMILES: COC1=CC(=CC=C1S(Cl)(=O)=O)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results