CAS RN 213261-59-7
CAS RN 213261-59-7
IUPAC Name:
(5-{5-[5-(hydroxymethyl)thiophen-2-yl]furan-2-yl}thiophen-2-yl)methanol
Synonyms:
(5-{5-[5-(hydroxymethyl)thiophen-2-yl]furan-2-yl}thiophen-2-yl)methanol
Molecular Weight (g/mol):
292.37
Molecular Formula:
C14H12O3S2
InChi Key:
KZENBFUSKMWCJF-UHFFFAOYSA-N
SMILES:
OCC1=CC=C(S1)C1=CC=C(O1)C1=CC=C(CO)S1
1
–
2
of
2
results
1
Thermo Scientific Chemicals RITA, p53 activator III
CAS: 213261-59-7 Molecular Formula: C14H12O3S2,C14H12O3S2 Synonym: 5,5′-(2,5-Furandiyl)bis-2-thiophenemethanol; NSC 652287
2
Tocris Bioscience™ RITA
CAS: 213261-59-7 Molecular Formula: C14H12O3S2