accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 213343-64-7

IUPAC Name: 1-[2-(2,5-diethylphospholan-1-yl)phenyl]-2,5-diethylphospholane cycloocta-1,5-diene tetrafluoroboranuide rhodium
Synonyms: 1,5-cyclooctadiene 1-[2-(2,5-diethylphospholan-1-yl)phenyl]-2,5-diethylphospholane rhodium tetrafluoroborate
Molecular Weight (g/mol): 660.37
Molecular Formula: C30H48BF4P2Rh
InChi Key: XIOPHSBHVOCZMM-UHFFFAOYNA-N
SMILES: [Rh].F[B-](F)(F)F.C1CC=CCCC=C1.CCC1CCC(CC)P1C1=CC=CC=C1P1C(CC)CCC1CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results