accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FBFFFPP(S)(S)(S)(S)H3CH3CCH3CH3Rh

CAS RN 213343-65-8

IUPAC Name: 1-[2-(2,5-dimethylphospholan-1-yl)ethyl]-2,5-dimethylphospholane cycloocta-1,5-diene tetrafluoroboranuide rhodium
Synonyms: 1,5-cyclooctadiene 1-[2-(2,5-dimethylphospholan-1-yl)ethyl]-2,5-dimethylphospholane rhodium tetrafluoroborate
Molecular Weight (g/mol): 556.22
Molecular Formula: C22H40BF4P2Rh
InChi Key: LKVIVYCYPYTYSO-UHFFFAOYNA-N
SMILES: [Rh].F[B-](F)(F)F.C1CC=CCCC=C1.CC1CCC(C)P1CCP1C(C)CCC1C
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results