accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClClClCl

CAS RN 2136-89-2

IUPAC Name: 1-chloro-2-(trichloromethyl)benzene
Synonyms: 2-chlorobenzotrichloride
Molecular Weight (g/mol): 229.91
Molecular Formula: C7H4Cl4
InChi Key: MFHPYLFZSCSNST-UHFFFAOYSA-N
SMILES: ClC1=CC=CC=C1C(Cl)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results