accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

N(R)N(R)O(S)(R)OOHNH2HOHOCH3

CAS RN 2140-61-6

IUPAC Name: 4-amino-1-[3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-methyl-1,2-dihydropyrimidin-2-one
Synonyms: 5-methylcytidine
Molecular Weight (g/mol): 257.25
Molecular Formula: C10H15N3O5
InChi Key: ZAYHVCMSTBRABG-UHFFFAOYNA-N
SMILES: CC1=CN(C2OC(CO)C(O)C2O)C(=O)N=C1N
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)