accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 21413-23-0

IUPAC Name: 3-chloro-2H-indazol-6-amine
Synonyms: 3-chloro-2H-indazol-6-amine
Molecular Weight (g/mol): 167.6
Molecular Formula: C7H6ClN3
InChi Key: DRRARKIFTNKQDW-UHFFFAOYSA-N
SMILES: NC1=CC2=NNC(Cl)=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results