accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COINOO

CAS RN 214279-40-0

IUPAC Name: 2-iodo-4-methoxy-1-nitrobenzene
Synonyms: 2-iodo-4-methoxy-1-nitrobenzene
Molecular Weight (g/mol): 279.03
Molecular Formula: C7H6INO3
InChi Key: NLTAHTYBZZSCIT-UHFFFAOYSA-N
SMILES: COC1=CC=C(C(I)=C1)[N+]([O-])=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results