accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3COBOH3CH3CNH2

CAS RN 214360-73-3

IUPAC Name: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Synonyms: 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Weight (g/mol): 219.09
Molecular Formula: C12H18BNO2
InChi Key: ZANPJXNYBVVNSD-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(N)C=C1
Quantity 
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)