accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3C

CAS RN 2146-37-4

IUPAC Name: ethylidenecyclopentane
Synonyms: cyclopentane, ethylidene-
Molecular Weight (g/mol): 96.17
Molecular Formula: C7H12
InChi Key: VONKRKBGTZDZNV-UHFFFAOYSA-N
SMILES: CC=C1CCCC1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results