accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2ClHOO

CAS RN 2148-56-3

IUPAC Name: 2-amino-6-chlorobenzoic acid
Synonyms: 2-amino-6-chlorobenzoic acid
Molecular Weight (g/mol): 171.58
Molecular Formula: C7H6ClNO2
InChi Key: SZCPTRGBOVXVCA-UHFFFAOYSA-N
SMILES: NC1=CC=CC(Cl)=C1C(O)=O
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results