accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3OONSH2N

CAS RN 214834-18-1

IUPAC Name: tert-butyl 4-carbamothioylpiperidine-1-carboxylate
Synonyms: tert-butyl 4-carbamothioylpiperidine-1-carboxylate
Molecular Weight (g/mol): 244.35
Molecular Formula: C11H20N2O2S
InChi Key: SCGQNJHAAYUQOO-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(CC1)C(N)=S
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results